ENA295068793 Enamine

ethyl 1-sulfamoylpyrrolidine-2-carboxylate

  • MDL number MFCD11649750
  • Cas Number : 2149055-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64515
Availability details for
Quantity
  • Availability
  • Add to Cart