ENA295068795 Enamine

3-methyl-2-(3-methylfuran-2-amido)benzoic acid

  • MDL number MFCD09045542
  • Cas Number : 926254-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-52732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-52732
Availability details for
Quantity
  • Availability
  • Add to Cart