ENA295068801 Enamine

N-[(thiophen-3-yl)methyl]pyridin-3-amine

  • MDL number MFCD11140123
  • Cas Number : 1019559-79-5
  • Molecular Weight : 190.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64523
Availability details for
Quantity
  • Availability
  • Add to Cart