ENA295068805 Enamine

1-(4-aminophenyl)-1H-pyrazole-3-carboxamide

  • MDL number MFCD09701488
  • Cas Number : 1154650-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64527
Availability details for
Quantity
  • Availability
  • Add to Cart