ENA295068807 Enamine

N-(2-methylphenyl)-1,2,3,4-tetrahydroquinoline-2-carboxamide

  • MDL number MFCD09944945
  • Cas Number : 1103844-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64530

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64530
Availability details for
Quantity
  • Availability
  • Add to Cart