ENA295068812 Enamine

2-oxo-1,2,5,6,7,8-hexahydroquinoline-3-carbonitrile

  • MDL number MFCD00971266
  • Cas Number : 4241-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 246...248
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53622

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53622
Availability details for
Quantity
  • Availability
  • Add to Cart