ENA295068816 Enamine

1-acetylpiperidine-4-carboxamide

  • MDL number MFCD02029734
  • Cas Number : 530120-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177...179
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64548

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64548
Availability details for
Quantity
  • Availability
  • Add to Cart