ENA295068817 Enamine

2-{4-[(methylcarbamoyl)methoxy]phenyl}acetic acid

  • MDL number MFCD11131697
  • Cas Number : 1019369-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64551
Availability details for
Quantity
  • Availability
  • Add to Cart