ENA295068820 Enamine

oct-7-en-4-one

  • MDL number MFCD11655177
  • Cas Number : 30503-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64554
Availability details for
Quantity
  • Availability
  • Add to Cart