ENA295068825 Enamine

1-[(4-bromophenyl)methyl]cyclohexan-1-amine

  • MDL number MFCD12153115
  • Cas Number : 1178611-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64562
Availability details for
Quantity
  • Availability
  • Add to Cart