ENA295068826 Enamine

2-amino-2-(pyridin-3-yl)acetic acid dihydrochloride

  • MDL number MFCD13586128
  • Cas Number : 1214125-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53927

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53927
Availability details for
Quantity
  • Availability
  • Add to Cart