ENA295068828 Enamine

3-(2-chlorophenyl)-1,2,4-oxadiazol-5-ol

  • MDL number MFCD16547699
  • Cas Number : 16672-15-4
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64566
Availability details for
Quantity
  • Availability
  • Add to Cart