ENA295068835 Enamine

3-(pyridine-3-carbonyl)-1H-indole

  • MDL number MFCD11211133
  • Cas Number : 37128-48-6
  • Molecular Weight : 222.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64579
Availability details for
Quantity
  • Availability
  • Add to Cart