ENA295068837 Enamine

methyl 3-chloro-4-(chlorosulfonyl)benzoate

  • MDL number MFCD06637330
  • Cas Number : 874830-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62464
Availability details for
Quantity
  • Availability
  • Add to Cart