ENA295068839 Enamine

4-(thiomorpholine-4-carbonyl)cyclohexan-1-amine

  • MDL number MFCD12163629
  • Cas Number : 1183736-33-5
  • Molecular Weight : 228.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64583
Availability details for
Quantity
  • Availability
  • Add to Cart