ENA295068844 Enamine

5-methyl-5-(5-methylfuran-2-yl)imidazolidine-2,4-dione

  • MDL number MFCD00568316
  • Cas Number : 485826-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62488

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62488
Availability details for
Quantity
  • Availability
  • Add to Cart