ENA295068847 Enamine

3-(pyridin-3-yl)-1,2,4-oxadiazol-5-amine

  • MDL number MFCD16301075
  • Cas Number : 49772-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64597
Availability details for
Quantity
  • Availability
  • Add to Cart