ENA295068848 Enamine

6-(2-chloroethoxy)-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD11164751
  • Cas Number : 58022-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64598
Availability details for
Quantity
  • Availability
  • Add to Cart