ENA295068850 Enamine

4-chloropyridine-3-sulfonyl chloride

  • MDL number MFCD09743982
  • Cas Number : 33263-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-64600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64600
Availability details for
Quantity
  • Availability
  • Add to Cart