ENA295068852 Enamine

4-bromo-N-[(4-hydroxyphenyl)methyl]-N-methyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD12849773
  • Cas Number : 1183454-77-4
  • Molecular Weight : 309.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64602

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64602
Availability details for
Quantity
  • Availability
  • Add to Cart