ENA295068863 Enamine

(2-aminoethyl)(methyl)(propan-2-yl)amine

  • MDL number MFCD09731337
  • Cas Number : 14157-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64614
Availability details for
Quantity
  • Availability
  • Add to Cart