ENA295068865 Enamine

2-[2-(1,3-dioxaindan-5-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD11540611
  • Cas Number : 1097005-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64618
Availability details for
Quantity
  • Availability
  • Add to Cart