ENA295068881 Enamine

N-(1-cyclopropylethyl)-N-methylaminosulfonamide

  • MDL number MFCD12194527
  • Cas Number : 1153065-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64652

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64652
Availability details for
Quantity
  • Availability
  • Add to Cart