ENA295068883 Enamine

2-{3-oxo-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-2-yl}acetonitrile

  • MDL number MFCD09944984
  • Cas Number : 1016878-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62533
Availability details for
Quantity
  • Availability
  • Add to Cart