ENA295068884 Enamine

2-(2,6-difluorophenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD07376377
  • Cas Number : 1017452-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64659
Availability details for
Quantity
  • Availability
  • Add to Cart