ENA295068888 Enamine

N-(4-fluorophenyl)-1,2,3,4-tetrahydroquinoline-2-carboxamide

  • MDL number MFCD09928655
  • Cas Number : 1103212-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64669
Availability details for
Quantity
  • Availability
  • Add to Cart