ENA295068896 Enamine

1-(azepan-1-yl)-2-hydroxyethan-1-one

  • MDL number MFCD08443510
  • Cas Number : 51068-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64693
Availability details for
Quantity
  • Availability
  • Add to Cart