ENA295068897 Enamine

{3-[(tert-butoxy)methyl]phenyl}methanamine

  • MDL number MFCD11196426
  • Cas Number : 538321-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64694
Availability details for
Quantity
  • Availability
  • Add to Cart