ENA295068899 Enamine

1-(1H-indazole-5-amido)cyclopentane-1-carboxylic acid

  • MDL number MFCD13671418
  • Cas Number : 1099147-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64698
Availability details for
Quantity
  • Availability
  • Add to Cart