ENA295068900 Enamine

6-chloroimidazo[2,1-b][1,3]thiazole-5-carbonitrile

  • MDL number MFCD00139631
  • Cas Number : 23576-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64700
Availability details for
Quantity
  • Availability
  • Add to Cart