ENA295068908 Enamine

3-(bromomethyl)-1-methyl-1,2-dihydroquinoxalin-2-one

  • MDL number MFCD13192552
  • Cas Number : 1501-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-182
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62617

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62617
Availability details for
Quantity
  • Availability
  • Add to Cart