ENA295068913 Enamine

3-methyl-5-(2-methylphenyl)-1H-1,2,4-triazole

  • MDL number MFCD11215058
  • Cas Number : 37894-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62626
Availability details for
Quantity
  • Availability
  • Add to Cart