ENA295068916 Enamine

3-sulfamoylbenzene-1-carbothioamide

  • MDL number MFCD09938063
  • Cas Number : 56542-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64735

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64735
Availability details for
Quantity
  • Availability
  • Add to Cart