ENA295068917 Enamine

3-[(1,3-thiazol-4-yl)methoxy]aniline

  • MDL number MFCD12823160
  • Cas Number : 1184653-06-2
  • Molecular Weight : 206.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(1,3-thiazol-4-ylmethoxy)aniline
Inchi Code 1S/C10H10N2OS/c11-8-2-1-3-10(4-8)13-5-9-6-14-7-12-9/h1-4,6-7H,5,11H2
Inchi Key IVIGYIAJQZNTTG-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62627
Availability details for
Quantity
  • Availability
  • Add to Cart