ENA295068921 Enamine

3-(methylsulfanyl)butan-1-amine

  • MDL number MFCD07786667
  • Cas Number : 887580-42-9
  • Molecular Weight : 119.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64743

Safety Information

signalword Danger
pcodes P210,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-64743
pictograms GHS05,GHS07
hstatements H227,H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart