ENA295068922 Enamine

3-amino-N-(4-bromo-2-methylphenyl)propanamide

  • MDL number MFCD09726273
  • Cas Number : 938517-29-4
  • Molecular Weight : 257.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-N-(4-bromo-2-methylphenyl)propanamide
Inchi Code 1S/C10H13BrN2O/c1-7-6-8(11)2-3-9(7)13-10(14)4-5-12/h2-3,6H,4-5,12H2,1H3,(H,13,14)
Inchi Key MEHJCODAEDSSJG-UHFFFAOYSA-N

Description

Purity 95
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-64744
Availability details for
Quantity
  • Availability
  • Add to Cart