ENA295068925 Enamine

3-[(2-hydroxy-4-methylphenyl)formamido]propanoic acid

  • MDL number MFCD09931846
  • Cas Number : 1016720-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64750
Availability details for
Quantity
  • Availability
  • Add to Cart