ENA295068942 Enamine

{3H-imidazo[4,5-b]pyridin-2-yl}methanol

  • MDL number MFCD05721720
  • Cas Number : 24638-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64780
Availability details for
Quantity
  • Availability
  • Add to Cart