ENA295068945 Enamine

1-amino-N-(prop-2-en-1-yl)cyclohexane-1-carboxamide

  • MDL number MFCD09929751
  • Cas Number : 1016691-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64794
Availability details for
Quantity
  • Availability
  • Add to Cart