ENA295068947 Enamine

3-(thiophen-3-yl)-1H-pyrazole

  • MDL number MFCD04035695
  • Cas Number : 19933-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64800

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64800
Availability details for
Quantity
  • Availability
  • Add to Cart