ENA295068953 Enamine

2-(aminomethyl)-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD16547734
  • Cas Number : 1240527-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64810
Availability details for
Quantity
  • Availability
  • Add to Cart