ENA295068963 Enamine

1-ethyl-4-isothiocyanato-1H-pyrazole

  • MDL number MFCD04969719
  • Cas Number : 1006471-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64839

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P285,P301+P312,P302+P352,P304+P340,P304+P341,P305+P351+P338,P310,P312,P330,P332+P313,P342+P311,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-64839
pictograms GHS05,GHS07,GHS08
hstatements H302,H315,H318,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart