ENA295068966 Enamine

6-(2-bromoacetyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD11182385
  • Cas Number : 58819-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64853

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64853
Availability details for
Quantity
  • Availability
  • Add to Cart