ENA295068970 Enamine

5-chloro-6-{[1-(pyridin-3-yl)ethyl]amino}pyridine-3-carboxylic acid

  • MDL number MFCD12679023
  • Cas Number : 1155521-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64862
Availability details for
Quantity
  • Availability
  • Add to Cart