ENA295068972 Enamine

4-methyl-3-(5-methyl-1H-1,2,3,4-tetrazol-1-yl)aniline

  • MDL number MFCD12091839
  • Cas Number : 1157100-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-64873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64873
Availability details for
Quantity
  • Availability
  • Add to Cart