ENA295068980 Enamine

(4-hydroxycyclohexyl)urea

  • MDL number MFCD12179633
  • Cas Number : 22457-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62648

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62648
Availability details for
Quantity
  • Availability
  • Add to Cart