ENA295068983 Enamine

N-[(2-methyl-1,3-thiazol-4-yl)methyl]pyridin-3-amine

  • MDL number MFCD11170613
  • Cas Number : 1042643-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64939

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64939
Availability details for
Quantity
  • Availability
  • Add to Cart