ENA295068985 Enamine

3-methylcyclohexane-1-thiol

  • MDL number MFCD10692150
  • Cas Number : 38699-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65030
Availability details for
Quantity
  • Availability
  • Add to Cart