ENA295068991 Enamine

1-(furan-3-carbonyl)-2,3-dihydro-1H-indol-5-amine

  • MDL number MFCD11137558
  • Cas Number : 1019597-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62657
Availability details for
Quantity
  • Availability
  • Add to Cart