ENA295068995 Enamine

1-(3-aminobenzoyl)piperidin-4-ol

  • MDL number MFCD09738889
  • Cas Number : 954264-64-3
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62660
Availability details for
Quantity
  • Availability
  • Add to Cart